Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172477.4 | NP_766065.1 | 12 - 13 | 2549 | 118 | |
GenBank mRNA | AK048041.1 | - | 12 - 13 | 2534 | 118 | |
AK076686.1 | - | 4 - 5 | 692 | 118 | ||
AK081176.1 | - | 12 - 13 | 2550 | 118 | ||
AK135781.1 | - | 10 - 11 | 1094 | 118 | ||
AK160784.1 | - | 12 - 13 | 2311 | 118 | ||
AK165202.1 | - | 12 - 13 | 2404 | 118 | ||
AK167298.1 | - | 12 - 13 | 2403 | 118 | ||
BC056367.1 | - | 12 - 13 | 2419 | 118 | ||
BC057310.1 | - | 12 - 13 | 2419 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |