Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170638.1 | NP_001164109.1 | 9 - 10 | 1145 | 72 | |
NM_009198.3 | NP_033224.3 | 9 - 10 | 1237 | 72 | ||
GenBank mRNA | AI663290.1 | - | 5 - 6 | 393 | 72 | |
AK164712.1 | - | 9 - 10 | 1147 | 72 | ||
BC013445.1 | - | 9 - 10 | 1178 | 4241557 | 72 | |
X77241.1 | - | 9 - 10 | 1104 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |