Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013491.2 | NP_038519.1 | 9 - 10 | 1151 | 68 | |
GenBank mRNA | AK036735.1 | - | 9 - 10 | 1153 | 68 | |
AK161489.1 | - | 2 - 3 | 116 | 68 | ||
AY046403.1 | - | 9 - 10 | 976 | 68 | ||
AY046404.1 | - | 10 - 11 | 1055 | 68 | ||
BC114336.1 | - | 9 - 10 | 1116 | 40048966 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |