Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl8a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289919.1 | NP_001276848.1 | 2 - 3 | 271 | 129 | |
NM_010088.2 | NP_034218.1 | 2 - 3 | 274 | 129 | ||
GenBank mRNA | AF011385.1 | - | 2 - 3 | 258 | 129 | |
AF015729.1 | - | 2 - 3 | 268 | 129 | ||
AK131668.1 | - | 2 - 3 | 272 | 129 | ||
AK146049.1 | - | 2 - 3 | 275 | 129 | ||
BC061150.1 | - | 2 - 3 | 271 | 30317747 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |