Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175484.3 | NP_780693.2 | 2 - 3 | 594 | 79 | |
GenBank mRNA | AK047330.1 | - | 2 - 3 | 511 | 79 | |
AK047527.1 | - | 2 - 3 | 536 | 79 | ||
AK087414.1 | - | 2 - 3 | 595 | 79 | ||
BC062649.1 | - | 2 - 3 | 326 | 6416110 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |