Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hmha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142701.1 | NP_001136173.1 | 14 - 15 | 1976 | 65 | |
NM_027521.3 | NP_081797.1 | 13 - 14 | 1528 | 65 | ||
GenBank mRNA | AK018130.1 | - | 1 - 2 | 43 | 65 | |
AK044860.1 | - | 13 - 14 | 1490 | 65 | ||
AK134029.1 | - | 7 - 8 | 950 | 65 | ||
AK151423.1 | - | 14 - 15 | 1920 | 65 | ||
AK159194.1 | - | 14 - 15 | 1977 | 65 | ||
AK161766.1 | - | 6 - 7 | 1036 | 65 | ||
AK170262.1 | - | 14 - 15 | 1962 | 65 | ||
AK171304.1 | - | 14 - 15 | 1962 | 65 | ||
BC053750.1 | - | 13 - 14 | 1528 | 6333971 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |