Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mier2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027422.2 | NP_081698.2 | 9 - 10 | 886 | 56 | |
GenBank mRNA | AK154672.1 | - | 9 - 10 | 903 | 56 | |
AK173128.1 | - | 8 - 9 | 1042 | 56 | ||
BC067013.1 | - | 9 - 10 | 886 | 6810181 | 56 | |
BC080757.1 | - | 2 - 3 | 137 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |