Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Susd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162913.1 | NP_001156385.1 | 2 - 3 | 301 | 69 | |
NM_027890.5 | NP_082166.3 | 4 - 5 | 661 | 69 | ||
GenBank mRNA | AK004703.2 | - | 4 - 5 | 631 | 69 | |
AK028817.1 | - | 4 - 5 | 637 | 69 | ||
AK033823.1 | - | 2 - 3 | 274 | 69 | ||
AK037059.1 | - | 4 - 5 | 643 | 69 | ||
AK037127.1 | - | 4 - 5 | 612 | 69 | ||
AK038944.1 | - | 4 - 5 | 662 | 69 | ||
AK042469.1 | - | 4 - 5 | 645 | 69 | ||
AK128959.1 | - | 4 - 5 | 633 | 69 | ||
AK167155.1 | - | 4 - 5 | 662 | 69 | ||
BC026837.1 | - | 4 - 5 | 614 | 4222578 | 69 | |
BC066833.1 | - | 2 - 3 | 279 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |