Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029781.3 | NP_084057.1 | 6 - 7 | 635 | 71 | |
GenBank mRNA | AB232632.1 | - | 5 - 6 | 390 | 71 | |
AK018269.1 | - | 6 - 7 | 654 | 71 | ||
AK038445.1 | - | 6 - 7 | 636 | 71 | ||
AK043946.1 | - | 6 - 7 | 659 | 71 | ||
AK046113.1 | - | 6 - 7 | 444 | 71 | ||
AK142025.1 | - | 5 - 6 | 1007 | 71 | ||
BC119094.1 | - | 6 - 7 | 623 | 8733843 | 71 | |
BC119098.1 | - | 6 - 7 | 610 | 8733847 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |