Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290633.1 | NP_001277562.1 | 9 - 10 | 1315 | 67 | |
NM_178256.4 | NP_839987.2 | 9 - 10 | 1318 | 67 | ||
GenBank mRNA | AK147432.1 | - | 6 - 7 | 597 | 67 | |
AY263368.1 | - | 9 - 10 | 1263 | 67 | ||
BC139012.1 | - | 9 - 10 | 1194 | 8862034 | 67 | |
BC145500.1 | - | 9 - 10 | 1191 | 9054063 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |