Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mocos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026779.1 | NP_081055.1 | 2 - 3 | 472 | 81 | |
GenBank mRNA | AK076385.1 | - | 2 - 3 | 286 | 81 | |
BC016223.1 | - | 2 - 3 | 287 | 81 | ||
BC113180.1 | - | 2 - 3 | 472 | 40060723 | 81 | |
BC113786.1 | - | 2 - 3 | 472 | 40060721 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |