Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144866.3 | NP_659115.3 | 4 - 5 | 588 | 155 | |
GenBank mRNA | AK049607.1 | - | 4 - 5 | 539 | 155 | |
AK077559.1 | - | 4 - 5 | 537 | 155 | ||
AK151028.1 | - | 4 - 5 | 489 | 155 | ||
AK151710.1 | - | 4 - 5 | 489 | 155 | ||
AK152428.1 | - | 4 - 5 | 489 | 155 | ||
AK152952.1 | - | 4 - 5 | 489 | 155 | ||
AK164020.1 | - | 4 - 5 | 539 | 155 | ||
AK166740.1 | - | 4 - 5 | 557 | 155 | ||
AK169345.1 | - | 4 - 5 | 561 | 155 | ||
AK170221.1 | - | 4 - 5 | 531 | 155 | ||
BC013717.1 | - | 4 - 5 | 515 | 3992883 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |