Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204914.1 | NP_001191843.1 | 6 - 7 | 724 | 68 | |
NM_144865.2 | NP_659114.2 | 6 - 7 | 730 | 68 | ||
GenBank mRNA | AK158694.1 | - | 1 - 2 | 304 | 68 | |
AY562230.1 | - | 6 - 7 | 563 | 68 | ||
BC020184.1 | - | 6 - 7 | 661 | 4505679 | 68 | |
EU555417.1 | - | 6 - 7 | 557 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |