Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009284.2 | NP_033310.2 | 17 - 18 | 2234 | 125 | |
GenBank mRNA | AF481808.1 | - | 16 - 17 | 1949 | 125 | |
AF481809.1 | - | 16 - 17 | 1949 | 125 | ||
AK149563.1 | - | 17 - 18 | 2194 | 126 | ||
AK163053.1 | - | 17 - 18 | 2235 | 125 | ||
AK206252.1 | - | 2 - 3 | 74 | 125 | ||
BC029318.1 | - | 4 - 5 | 423 | 125 | ||
BC127054.1 | - | 17 - 18 | 2024 | 40109712 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |