Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl8a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271378.1 | NP_001258307.1 | 2 - 3 | 314 | 227 | |
NM_001271379.1 | NP_001258308.1 | 2 - 3 | 314 | 122 | ||
NM_011167.3 | NP_035297.1 | 2 - 3 | 311 | 227 | ||
NR_073166.1 | 2 - 3 | 236 | 227 | |||
GenBank mRNA | AF090140.1 | - | 2 - 3 | 220 | 227 | |
AK005435.1 | - | 2 - 3 | 275 | 227 | ||
AK005478.1 | - | 2 - 3 | 315 | 122 | ||
BC099469.1 | - | 2 - 3 | 257 | 30787452 | 227 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |