Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slco2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252530.1 | NP_001239459.1 | 10 - 11 | 1909 | 91 | |
NM_001252531.1 | NP_001239460.1 | 10 - 11 | 1852 | 91 | ||
NM_175316.3 | NP_780525.1 | 10 - 11 | 1655 | 91 | ||
GenBank mRNA | AK033539.1 | - | 2 - 3 | 281 | 91 | |
AK041144.1 | - | 10 - 11 | 1851 | 91 | ||
AK048120.1 | - | 10 - 11 | 1656 | 91 | ||
AK087682.1 | - | 10 - 11 | 1907 | 91 | ||
AK132394.1 | - | 10 - 11 | 1910 | 91 | ||
AK163542.1 | - | 10 - 11 | 1905 | 91 | ||
AK173049.1 | - | 8 - 9 | 1441 | 91 | ||
BC019209.1 | - | 10 - 11 | 1638 | 91 | ||
BC037089.1 | - | 10 - 11 | 1627 | 91 | ||
BC049251.1 | - | 10 - 11 | 1618 | 91 | ||
BC096485.1 | - | 10 - 11 | 1664 | 5101188 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |