Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010832.5 | NP_034962.2 | 5 - 6 | 592 | 129 | |
GenBank mRNA | AF117066.2 | - | 5 - 6 | 594 | 129 | |
AK089368.1 | - | 5 - 6 | 675 | 129 | ||
AK148898.1 | - | 5 - 6 | 570 | 129 | ||
BC010226.1 | - | 5 - 6 | 553 | 3501126 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |