Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Prl2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025532.3 | NP_079808.1 | 3 - 4 | 391 | 117 | |
GenBank mRNA | AF525155.1 | - | 3 - 4 | 342 | 117 | |
AK005411.1 | - | 3 - 4 | 392 | 117 | ||
AK009863.1 | - | 3 - 4 | 391 | 117 | ||
BC100329.1 | - | 3 - 4 | 381 | 30787440 | 117 | |
BC111112.1 | - | 3 - 4 | 387 | 40050224 | 117 | |
BC138199.1 | - | 3 - 4 | 384 | 8861218 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |