Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Podxl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013723.3 | NP_038751.2 | 6 - 7 | 1439 | 64 | |
GenBank mRNA | AB028048.1 | - | 6 - 7 | 1320 | 64 | |
AF290209.1 | - | 6 - 7 | 1338 | 64 | ||
AK134723.1 | - | 2 - 3 | 175 | 64 | ||
BC052442.1 | - | 6 - 7 | 1413 | 6406610 | 64 | |
BC054530.1 | - | 6 - 7 | 1413 | 6406610 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |