Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl21b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205036.1 | NP_001191965.1 | 1 - 2 | 174 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |