Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144537.3 | NP_653120.1 | 10 - 11 | 1565 | 65 | |
GenBank mRNA | AF466283.1 | - | 10 - 11 | 1566 | 65 | |
AK015256.1 | - | 10 - 11 | 1568 | 65 | ||
BC138114.1 | - | 9 - 10 | 1083 | 8861133 | 65 | |
BC145151.1 | - | 9 - 10 | 950 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |