Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Adamts14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081127.1 | NP_001074596.1 | 15 - 16 | 2425 | 76 | |
GenBank mRNA | AK145103.1 | - | 15 - 16 | 2427 | 76 | |
BC157953.1 | - | 15 - 16 | 2260 | 9007283 | 76 | |
BC158014.1 | - | 15 - 16 | 2258 | 9088151 | 76 | |
BC172106.1 | - | 15 - 16 | 2241 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |