Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sipa1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081028.1 | NP_001074497.1 | 19 - 20 | 5442 | 92 | |
GenBank mRNA | AK012129.1 | - | 2 - 3 | 413 | 92 | |
AK044193.1 | - | 6 - 7 | 1269 | 92 | ||
AK087819.1 | - | 6 - 7 | 1427 | 92 | ||
AK144537.1 | - | 4 - 5 | 686 | 92 | ||
AK172968.1 | - | 13 - 14 | 2281 | 92 | ||
BC034106.1 | - | 2 - 3 | 266 | 92 | ||
BC095969.1 | - | 9 - 10 | 1972 | 92 | ||
BC116408.1 | - | 4 - 5 | 1035 | 92 | ||
BC128493.1 | - | 4 - 5 | 1032 | 92 | ||
BC144792.1 | - | 6 - 7 | 1345 | 92 | ||
BC145739.1 | - | 6 - 7 | 1345 | 92 | ||
BC152360.1 | - | 4 - 5 | 1032 | 92 | ||
BC152361.1 | - | 4 - 5 | 1020 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |