Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mypn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182992.2 | NP_892037.2 | 16 - 17 | 3506 | 148 | |
GenBank mRNA | AK220521.1 | - | 16 - 17 | 3506 | 148 | |
BC052872.1 | - | 2 - 3 | 174 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |