Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172290.3 | NP_758494.2 | 5 - 6 | 862 | 67 | |
GenBank mRNA | AK045973.1 | - | 5 - 6 | 862 | 67 | |
AK046377.1 | - | 5 - 6 | 862 | 67 | ||
AK158752.1 | - | 4 - 5 | 881 | 67 | ||
BC023307.1 | - | 4 - 5 | 702 | 4480983 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |