Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010140.3 | NP_034270.1 | 5 - 6 | 1420 | 79 | |
GenBank mRNA | AK041935.1 | - | 5 - 6 | 1421 | 79 | |
AK045219.1 | - | 5 - 6 | 1421 | 79 | ||
AK084921.1 | - | 5 - 6 | 1400 | 79 | ||
AK132335.1 | - | 5 - 6 | 1369 | 79 | ||
BC053479.1 | - | 1 - 2 | 312 | 79 | ||
BC060164.1 | - | 5 - 6 | 1397 | 79 | ||
BC093483.1 | - | 1421 | 79 | |||
BC138641.1 | - | 5 - 6 | 1343 | 8861661 | 79 | |
BC138643.1 | - | 5 - 6 | 1343 | 8861663 | 79 | |
BC145349.1 | - | 5 - 6 | 1343 | 9053905 | 79 | |
M68513.1 | - | 5 - 6 | 1395 | 79 | ||
M68515.1 | - | 5 - 6 | 1395 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |