Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053176.2 | NP_444406.2 | 5 - 6 | 715 | 136 | |
GenBank mRNA | AB055897.1 | - | 5 - 6 | 673 | 136 | |
AF194028.1 | - | 5 - 6 | 655 | 136 | ||
AY135662.1 | - | 5 - 6 | 676 | 136 | ||
BC011168.1 | - | 5 - 6 | 661 | 4195766 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |