Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078167.2 | NP_001071635.1 | 2 - 3 | 840 | 65 | |
NM_173374.4 | NP_775550.2 | 2 - 3 | 840 | 65 | ||
GenBank mRNA | AK053674.1 | - | 2 - 3 | 531 | 65 | |
AK078715.1 | - | 2 - 3 | 497 | 65 | ||
AK150535.1 | - | 2 - 3 | 543 | 65 | ||
AK169203.1 | - | 2 - 3 | 497 | 65 | ||
AK169623.1 | - | 2 - 3 | 498 | 65 | ||
BC046773.1 | - | 2 - 3 | 472 | 5708288 | 65 | |
BC051213.1 | - | 2 - 3 | 65 | |||
BC058627.1 | - | 2 - 3 | 506 | 6401674 | 65 | |
X66091.1 | - | 2 - 3 | 453 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |