Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122603.1 | NP_001116075.1 | 17 - 18 | 7107 | 62 | |
GenBank mRNA | AK040441.1 | - | 7 - 8 | 3811 | 62 | |
BC024520.1 | - | 2 - 3 | 769 | 62 | ||
BC024554.1 | - | 2 - 3 | 62 | |||
BC026653.1 | - | 3 - 4 | 1312 | 62 | ||
BC029250.1 | - | 5 - 6 | 2129 | 62 | ||
BC030871.1 | - | 6 - 7 | 2956 | 62 | ||
BC036158.1 | - | 3 - 4 | 1149 | 62 | ||
BC036159.1 | - | 2 - 3 | 431 | 62 | ||
BC039286.1 | - | 8 - 9 | 3533 | 62 | ||
BC048952.1 | - | 1 - 2 | 303 | 62 | ||
BC055475.1 | - | 3 - 4 | 820 | 62 | ||
BC069956.1 | - | 3 - 4 | 1115 | 62 | ||
BC092067.1 | - | 3 - 4 | 1186 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |