Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194263.2 | NP_919239.1 | 7 - 8 | 1452 | 113 | |
GenBank mRNA | AF306667.1 | - | 7 - 8 | 1390 | 113 | |
AK052389.1 | - | 7 - 8 | 1435 | 113 | ||
AK141251.1 | - | 7 - 8 | 1222 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |