Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019493.3 | NP_062366.2 | 2 - 3 | 398 | 92 | |
GenBank mRNA | AB050983.2 | - | 2 - 3 | 398 | 92 | |
AJ005120.1 | - | 2 - 3 | 271 | 92 | ||
AK136041.1 | - | 2 - 3 | 385 | 92 | ||
AK136124.1 | - | 2 - 3 | 386 | 92 | ||
AK136149.1 | - | 2 - 3 | 388 | 92 | ||
AK136185.1 | - | 2 - 3 | 390 | 92 | ||
AK161379.1 | - | 4 - 5 | 477 | 92 | ||
AK162122.1 | - | 2 - 3 | 385 | 92 | ||
AK163407.1 | - | 2 - 3 | 384 | 92 | ||
BC066811.1 | - | 2 - 3 | 372 | 30475849 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |