Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dixdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178118.2 | NP_835219.1 | 5 - 6 | 874 | 76 | |
GenBank mRNA | AK129432.1 | - | 1 - 2 | 54 | 76 | |
AK164424.1 | - | 5 - 6 | 797 | 76 | ||
AY549883.1 | - | 5 - 6 | 732 | 76 | ||
AY549884.1 | - | 5 - 6 | 735 | 76 | ||
BC048182.1 | - | 5 - 6 | 865 | 6826299 | 76 | |
BC063085.1 | - | 5 - 6 | 865 | 6826299 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |