Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Chordc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025844.2 | NP_080120.1 | 7 - 8 | 688 | 151 | |
GenBank mRNA | AK003259.1 | - | 7 - 8 | 689 | 151 | |
AK132696.1 | - | 668 | 151 | |||
BC018374.1 | - | 7 - 8 | 688 | 4236535 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |