Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172495.5 | NP_766083.3 | 5 - 6 | 807 | 146 | |
GenBank mRNA | AK128935.1 | - | 4 - 5 | 630 | 146 | |
AK144324.1 | - | 5 - 6 | 639 | 146 | ||
BC076623.1 | - | 3 - 4 | 413 | 146 | ||
BC141150.1 | - | 4 - 5 | 541 | 9055762 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |