Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168304.1 | NP_001161776.1 | 5 - 6 | 800 | 75 | |
NM_001291816.1 | NP_001278745.1 | 4 - 5 | 724 | 75 | ||
GenBank mRNA | AK052818.1 | - | 5 - 6 | 631 | 75 | |
AK122421.1 | - | 5 - 6 | 800 | 75 | ||
AK147340.1 | - | 4 - 5 | 522 | 75 | ||
BC048908.1 | - | 5 - 6 | 550 | 75 | ||
BC080280.1 | - | 4 - 5 | 674 | 75 | ||
BC137614.1 | - | 5 - 6 | 754 | 8860633 | 75 | |
BC144782.1 | - | 5 - 6 | 754 | 9053330 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |