Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256432.1 | NP_001243361.1 | 1 - 2 | 197 | 123 | |
NM_177671.5 | NP_808339.2 | 1 - 2 | 197 | 123 | ||
GenBank mRNA | AK039700.1 | - | 1 - 2 | 160 | 123 | |
AK143168.1 | - | 1082 | 123 | |||
BC116991.1 | - | 1 - 2 | 127 | 40126310 | 123 | |
BC119084.1 | - | 1 - 2 | 127 | 8733833 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |