Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thumpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008188.2 | NP_032214.1 | 6 - 7 | 1113 | 77 | |
GenBank mRNA | AK142140.1 | - | 6 - 7 | 1132 | 77 | |
BC046800.1 | - | 6 - 7 | 1113 | 6403880 | 77 | |
U83176.1 | - | 6 - 7 | 1107 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |