Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.1 ST Array |
10565600 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0700031630.mm.1 |
NM_001177412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031630.mm.1 |
NM_001177412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031630.mm.1 |
TC0700003763.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031633.mm.1 |
NM_001177412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031633.mm.1 |
NM_001177412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031633.mm.1 |
TC0700003763.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031631.mm.1 |
NM_001177412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031631.mm.1 |
NM_001177412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031631.mm.1 |
TC0700003763.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031636.mm.1 |
NM_001177412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031636.mm.1 |
NM_001177412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031636.mm.1 |
TC0700003763.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031635.mm.1 |
NM_001177412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031635.mm.1 |
NM_001177412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031635.mm.1 |
TC0700003763.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031638.mm.1 |
NM_001177412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031638.mm.1 |
NM_001177412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031638.mm.1 |
TC0700003763.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031627.mm.1 |
NM_001177412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031627.mm.1 |
NM_001177412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031627.mm.1 |
TC0700003763.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031628.mm.1 |
NM_001177412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031628.mm.1 |
NM_001177412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031628.mm.1 |
TC0700003763.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031637.mm.1 |
NM_001177412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031637.mm.1 |
NM_001177412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031637.mm.1 |
TC0700003763.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031634.mm.1 |
NM_001177412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031634.mm.1 |
NM_001177412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031634.mm.1 |
TC0700003763.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031639.mm.1 |
NM_001177412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031639.mm.1 |
NM_001177412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031639.mm.1 |
TC0700003763.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031640.mm.1 |
NM_001177412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031640.mm.1 |
NM_001177412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031640.mm.1 |
TC0700003763.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031629.mm.1 |
NM_001177412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031629.mm.1 |
NM_001177412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031629.mm.1 |
TC0700003763.mm.1 |
MTA TC |
Murine Genome U74v2 Set |
133191_at |
- |
- |
Murine Genome U74v2 Set |
133191_PM_at |
- |
- |