Display Info New header:
tfpt-header-footer.tfcom-eks-na-comerce-gateway-prd.commerceprod.thermofisher.net cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@2ef2c419
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:/anonymous enableSimpleHeaderValue:
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Gene Expression
Search All
Popular Product Areas
TaqMan Assays
Gene Expression
Copy Number Variation
Mutation Detection
miRNA
SNP Genotyping
Primary Antibodies
Secondary Antibodies
ELISA Kits
All Documents & Support
Certificates
SDS
Manuals & Protocols
Product FAQs
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm01183330_g1
See other KLRK1 GE Assays ›
Gene Symbol:
Klrk1
Gene Name:
killer cell lectin-like receptor subfamily K, member 1
Gene Aliases:
D6H12S2489E, NKG2-D, Nkg2d
Chromosome Location:
Chr.6: 129610323 - 129623864 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID
Mm01183330_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
Displaying...
Build 38.1 Mouse Chr.6: Mm01183330_g1
×
Created with Raphaël 2.1.2
Klrk1
NM_03307
8.4
NM_00108
3322.2
AK170499
.1
BC057147
.1
BY207066
.1
129,568,
000
129,569,
000
129,570,
000
129,571,
000
129,572,
000
129,573,
000
129,574,
000
129,575,
000
129,576,
000
129,577,
000
129,578,
000
129,579,
000
129,580,
000
129,581,
000
129,582,
000
129,583,
000
129,584,
000
129,585,
000
129,586,
000
129,587,
000
129,588,
000
129,589,
000
129,590,
000
129,591,
000
129,592,
000
129,593,
000
129,594,
000
129,595,
000
129,596,
000
129,597,
000
129,598,
000
129,599,
000
129,600,
000
129,601,
000
129,602,
000
129,603,
000
129,604,
000
129,605,
000
129,606,
000
129,607,
000
129,608,
000
129,609,
000
129,610,
000
129,611,
000
129,612,
000
129,613,
000
129,614,
000
129,615,
000
129,616,
000
129,617,
000
129,618,
000
129,619,
000
129,620,
000
129,621,
000
129,622,
000
129,623,
000
129,624,
000
129,625,
000
129,626,
000
129,627,
000
129,628,
000
129,629,
000
129,630,
000
129,631,
000
129,632,
000
129,633,
000
129,634,
000
129,635,
000
129,636,
000
129,637,
000
129,638,
000
129,639,
000
129,640,
000
129,641,
000
129,642,
000
129,643,
000
129,644,
000
129,645,
000
129,646,
000
129,647,
000
129,648,
000
129,649,
000
129,650,
000
129,651,
000
129,652,
000
129,653,
000
129,654,
000
129,655,
000
129,656,
000
129,657,
000
129,658,
000
129,659,
000
129,660,
000
129,661,
000
129,662,
000
129,663,
000
129,664,
000
129,665,
000
129,666,
000
Show all RefSeq and GenBank
Back To Top
Assay Details
Gene Symbol: Klrk1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001083322.2
NP_001076791.1
4 - 5
301
75
NM_033078.4
NP_149069.1
4 - 5
394
75
GenBank mRNA
AF030313.1
-
4 - 5
278
75
AF039026.1
-
4 - 5
308
75
AF054819.1
-
5 - 6
568
75
AK040436.1
-
4 - 5
383
75
BC057147.1
-
4 - 5
345
5328432
75
CT010384.1
-
4 - 5
284
75
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as MTA transcript cluster(s) TC0600003231.mm.1
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Mammalian Gene Collection
Detect Genomic DNA
Mus musculus
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Klrk1
Gene Ontology Categories:
Function(s)
Process(es)
stimulatory C-type lectin receptor signaling pathway
immune system process
positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
natural killer cell activation
cell differentiation
positive regulation of myeloid dendritic cell activation
positive regulation of interferon-gamma production
positive regulation of apoptotic process
innate immune response
positive regulation of nitric oxide biosynthetic process
positive regulation of natural killer cell mediated cytotoxicity
defense response to Gram-positive bacterium
cellular response to lipopolysaccharide
protein binding
kinase binding
carbohydrate binding
MHC class Ib receptor activity
MHC class I protein binding
protein homodimerization activity
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Mouse Clariom S Array
PSR0600025253.mm.1
TC0600003231.mm.1
MTA TC
Affymetrix Mouse Clariom S Array
PSR0600025253.mm.1
NM_001083322.2
Refseq
Affymetrix Mouse Clariom S Array
PSR0600025253.mm.1
NM_001286018.1
Refseq
Affymetrix Mouse Clariom S Array
PSR0600025253.mm.1
NM_033078.4
Refseq
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:/anonymous
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c659bcd8-d8sjm:80/100.66.76.197:80.
git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-Offline