Search Thermo Fisher Scientific
Gene Symbol: Slc2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011401.4 | NP_035531.3 | 8 - 9 | 1420 | 77 | |
GenBank mRNA | AK028041.1 | - | 8 - 9 | 1196 | 77 | |
AK045328.1 | - | 8 - 9 | 1196 | 77 | ||
AK147258.1 | - | 7 - 8 | 3732 | 77 | ||
AK155997.1 | - | 8 - 9 | 1179 | 77 | ||
AK164286.1 | - | 8 - 9 | 1192 | 77 | ||
BC034122.1 | - | 8 - 9 | 1178 | 4527667 | 77 | |
BC050095.1 | - | 8 - 9 | 1174 | 77 | ||
BC058811.1 | - | 8 - 9 | 1178 | 6490844 | 77 | |
CT010354.1 | - | 7 - 8 | 1065 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |