Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tuba8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017379.1 | NP_059075.1 | 2 - 3 | 296 | 69 | |
GenBank mRNA | AJ245923.1 | - | 2 - 3 | 296 | 69 | |
AK032157.1 | - | 269 | 69 | |||
BC017631.1 | - | 2 - 3 | 275 | 4507364 | 69 | |
GU591980.1 | - | 248 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |