Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC048546 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001179.3 | NP_001001179.2 | 26 - 27 | 3361 | 77 | |
GenBank mRNA | AK138836.1 | - | 26 - 27 | 3362 | 77 | |
AK161184.1 | - | 26 - 27 | 3365 | 77 | ||
BC034132.1 | - | 1 - 2 | 121 | 77 | ||
BC040781.1 | - | 14 - 15 | 1813 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |