Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tead4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080979.1 | NP_001074448.2 | 6 - 7 | 1002 | 84 | |
NM_011567.2 | NP_035697.3 | 7 - 8 | 1131 | 84 | ||
GenBank mRNA | AB221623.1 | - | 5 - 6 | 432 | 83 | |
AK133915.1 | - | 7 - 8 | 792 | 84 | ||
BC130257.1 | - | 5 - 6 | 437 | 40142595 | 84 | |
D87965.1 | - | 6 - 7 | 1002 | 84 | ||
D87966.1 | - | 7 - 8 | 1126 | 84 | ||
L26343.1 | - | 7 - 8 | 671 | 84 | ||
L26344.1 | - | 616 | 84 | |||
U51743.1 | - | 5 - 6 | 417 | 84 | ||
X94441.1 | - | 6 - 7 | 562 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |