Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Morc2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159288.1 | NP_001152760.1 | 10 - 11 | 1551 | 73 | |
NM_198162.1 | NP_937805.1 | 9 - 10 | 878 | 73 | ||
GenBank mRNA | AK173042.1 | - | 10 - 11 | 1551 | 73 | |
BC059243.1 | - | 9 - 10 | 878 | 6417551 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |