Search Thermo Fisher Scientific
Gene Symbol: Fam50a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138607.3 | NP_613073.2 | 7 - 8 | 828 | 126 | |
GenBank mRNA | AK008402.1 | - | 7 - 8 | 829 | 126 | |
BC026128.1 | - | 6 - 7 | 1201 | 126 | ||
BC125297.1 | - | 6 - 7 | 694 | 40129812 | 126 | |
Y18505.1 | - | 7 - 8 | 793 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |