Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016981.2 | NP_058677.1 | 8 - 9 | 2599 | 88 | |
GenBank mRNA | AK150135.1 | - | 8 - 9 | 2602 | 88 | |
AK154857.1 | - | 8 - 9 | 2636 | 88 | ||
BC051431.1 | - | 8 - 9 | 1946 | 6529306 | 88 | |
BC052708.1 | - | 8 - 9 | 2481 | 5702902 | 88 | |
U51112.1 | - | 8 - 9 | 2142 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |