Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Katna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011835.2 | NP_035965.2 | 3 - 4 | 451 | 68 | |
GenBank mRNA | AF153197.1 | - | 3 - 4 | 429 | 68 | |
AK012319.1 | - | 3 - 4 | 451 | 68 | ||
AK041900.1 | - | 3 - 4 | 446 | 68 | ||
AK208851.1 | - | 2 - 3 | 68 | |||
BC009136.1 | - | 3 - 4 | 365 | 3597742 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |