Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007939.2 | NP_031965.2 | 10 - 11 | 1993 | 66 | |
GenBank mRNA | AK220393.1 | - | 8 - 9 | 1477 | 66 | |
U72207.1 | - | 10 - 11 | 2002 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Detect Genomic DNA Mus musculus Probe spans exons |