Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tecpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027410.1 | NP_081686.1 | 13 - 14 | 2238 | 80 | |
GenBank mRNA | AB093295.1 | - | 12 - 13 | 2407 | 80 | |
AK032858.1 | - | 13 - 14 | 2212 | 80 | ||
AK169888.1 | - | 13 - 14 | 2185 | 80 | ||
AK170108.1 | - | 13 - 14 | 2185 | 80 | ||
AK186297.1 | - | 1 - 2 | 146 | 80 | ||
BC046968.1 | - | 13 - 14 | 2238 | 6306206 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |