Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pyroxd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029011.2 | NP_083287.2 | 3 - 4 | 558 | 65 | |
GenBank mRNA | AK038414.1 | - | 3 - 4 | 559 | 65 | |
AK154222.1 | - | 3 - 4 | 320 | 65 | ||
AK160239.1 | - | 3 - 4 | 274 | 65 | ||
BC140971.1 | - | 3 - 4 | 387 | 9055583 | 65 | |
BC144951.1 | - | 3 - 4 | 387 | 9053499 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |